Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chromyl chloride

Catalog No. 20004210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-004-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-004-210G Supplier Sigma Aldrich Supplier No. 20004210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chromyl chloride

Specifications

CAS 14977-61-8
Melting Point -96.5°C
Density 1.911 g/mL (at 25°C)
Boiling Point 117°C
Molecular Formula Cl2CrO2
Linear Formula CrO2Cl2
Quantity 10 g
MDL Number MFCD00010951
Synonym Chromium(VI) dichloride dioxide; Chromium(VI) oxychloride; Dichlorochromium(VI) oxide
Molecular Weight (g/mol) 154.9
Percent Purity ≥99.99% trace metals basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.