CAS RN 149817-62-9
IUPAC Name:
4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine
Synonyms:
4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular Weight (g/mol):
312.17
Molecular Formula:
C15H10BrN3
InChi Key:
CJRRILXBSCRHKN-UHFFFAOYSA-N
SMILES:
BrC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
1
–
1
of
1
results
1
4'-Bromo-2,2':6',2″-terpyridine 97.0+%, TCI America™
CAS: 149817-62-9 Molecular Formula: C15H10BrN3 Molecular Weight (g/mol): 312.17 MDL Number: MFCD06796965 InChI Key: CJRRILXBSCRHKN-UHFFFAOYSA-N PubChem CID: 10614995 IUPAC Name: 4-bromo-6-(pyridin-2-yl)-2,2'-bipyridine SMILES: BrC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1