accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1498333-89-3

IUPAC Name: (9H-fluoren-9-yl)methyl N-[(pyridin-3-yl)methyl]carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-(pyridin-3-ylmethyl)carbamate
Molecular Weight (g/mol): 330.39
Molecular Formula: C21H18N2O2
InChi Key: HIELIWLCJOQJFQ-UHFFFAOYSA-N
SMILES: O=C(NCC1=CN=CC=C1)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results