Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Fmoc-aminomethyl)pyridine, 97%, Thermo Scientific™

Catalog No. AAH6687506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6687506 5 g
AAH6687503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6687506 Supplier Thermo Scientific Chemicals Supplier No. H6687506

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1498333-89-3
Molecular Formula C21H18N2O2
Molecular Weight (g/mol) 330.387
InChI Key HIELIWLCJOQJFQ-UHFFFAOYSA-N
Synonym 9h-fluoren-9-ylmethyl n-pyridin-3-ylmethyl carbamate, 9h-fluoren-9-yl methyl pyridin-3-ylmethyl carbamate
PubChem CID 86277479
IUPAC Name 9H-fluoren-9-ylmethyl N-(pyridin-3-ylmethyl)carbamate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCC4=CN=CC=C4

Specifications

Color White
Quantity 5 g
Formula Weight 330.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(Fmoc-aminomethyl)pyridine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.