accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14998-27-7

IUPAC Name: chlorite
Synonyms: chlorite
Molecular Weight (g/mol): 67.45
Molecular Formula: ClO2
InChi Key: QBWCMBCROVPCKQ-UHFFFAOYSA-M
SMILES: [O-][Cl]=O
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results