Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPH Aliphatic Hydrocarbon Standard, 1000μg/mL in water, Ultra Scientific

Catalog No. USICC012
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICC012 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICC012 Supplier Agilent Technologies Supplier No. ICC012
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14998-27-7 , 7732-18-5
Molecular Formula ClO2-
Molecular Weight (g/mol) 67.448
InChI Key QBWCMBCROVPCKQ-UHFFFAOYSA-M
PubChem CID 197148
ChEBI CHEBI:17441
IUPAC Name chlorite
SMILES [O-]Cl=O

Specifications

CAS Max % 99.9, 0.1
Analyte or Component Names Water,chlorite,soluble compounds as ClO2
Element Chlorite, Soluble compounds as ClO2
Solvent or Matrix Water
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material EPH Aliphatic Hydrocarbon Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.