accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14999-52-1

IUPAC Name: disodium [(2R,3S,5R)-3-hydroxy-5-(6-oxo-6,9-dihydro-1H-purin-9-yl)oxolan-2-yl]methyl phosphate
Synonyms: disodium dIMP
Molecular Weight (g/mol): 376.17
Molecular Formula: C10H11N4Na2O7P
InChi Key: VDPLPYMIJCLJEF-DZVPOJRENA-L
SMILES: [Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=NC2=C1N=CNC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results