accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 150-19-6

IUPAC Name: 3-methoxyphenol
Synonyms: 3-methoxyphenol M-methoxyphenol m-guaiacol M-hydroxyanisole
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: ASHGTJPOSUFTGB-UHFFFAOYSA-N
SMILES: COC1=CC=CC(O)=C1
Boiling Point 
  • (2)
  • (6)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (2)
  • (6)
  • (3)
  • (6)
  • (5)
  • (3)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (6)