accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SPSH3CSCH3

CAS RN 150-50-5

IUPAC Name: tris(butylsulfanyl)phosphane
Synonyms: merphos
Molecular Weight (g/mol): 298.5
Molecular Formula: C12H27PS3
InChi Key: KLAPGAOQRZTCBI-UHFFFAOYSA-N
SMILES: CCCCSP(SCCCC)SCCCC
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)