accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 150-96-9

IUPAC Name: 3-hydroxy-3-methylpentanoic acid
Synonyms: 3-hydroxy-3-methylpentanoic acid
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: KEGHVPSZIWXTPY-UHFFFAOYNA-N
SMILES: CCC(C)(O)CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results