Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-3-methylvaleric Acid 95.0+%, TCI America™
SDP

Catalog No. H06265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06265G 5 g
H062625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06265G Supplier TCI America Supplier No. H06265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 150-96-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00059657
InChI Key KEGHVPSZIWXTPY-UHFFFAOYSA-N
Synonym 3-Hydroxy-3-methylpentanoic Acid
PubChem CID 538288
IUPAC Name 3-hydroxy-3-methylpentanoic acid
SMILES CCC(C)(CC(=O)O)O

Specifications

Color Yellow
Boiling Point 144°C
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Hydroxy-3-methylvaleric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.