accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNHN

CAS RN 1500-85-2

IUPAC Name: 7H-pyrrolo[2,3-d]pyrimidin-4-amine
Synonyms: 7-deazaadenine
Molecular Weight (g/mol): 134.14
Molecular Formula: C6H6N4
InChi Key: PEHVGBZKEYRQSX-UHFFFAOYSA-N
SMILES: NC1=C2C=CNC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results