CAS RN 150080-09-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Val-boroPro
An orally available amino boronic acid dipeptide with antihyperglycemic and anticancer properties
2