accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHO

CAS RN 1501-27-5

IUPAC Name: 5-methoxy-5-oxopentanoic acid
Synonyms: glutaric acid monomethyl ester glutaric acid methyl half ester methyl glutarate,mono
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: IBMRTYCHDPMBFN-UHFFFAOYSA-N
SMILES: COC(=O)CCCC(O)=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results