Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl Glutarate 97.0+%, TCI America™
SDP

Catalog No. G025825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G025825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G025825G Supplier TCI America Supplier No. G025825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1501-27-5
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00004409
InChI Key IBMRTYCHDPMBFN-UHFFFAOYSA-N
Synonym monomethyl glutarate, methyl hydrogen glutarate, mono-methyl glutarate, glutaric acid monomethyl ester, monomethyl glutaric acid, 4-methoxycarbonylbutanoic acid, glutaric acid, monomethyl ester, pentanedioic acid, monomethyl ester, 4-methoxycarbonyl butyric acid, glutaric acid methyl half ester
PubChem CID 73917
ChEBI CHEBI:86396
IUPAC Name 5-methoxy-5-oxopentanoic acid
SMILES COC(=O)CCCC(=O)O

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 146.14
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Monomethyl Glutarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.