accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1501-82-2

IUPAC Name: (Z)-cyclododecene
Synonyms: (Z)-cyclododecene
Molecular Weight (g/mol): 166.31
Molecular Formula: C12H22
InChi Key: HYPABJGVBDSCIT-UPHRSURJSA-N
SMILES: C1CCCCC\C=C/CCCC1
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)