Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclododecene, ca 70% trans isomer, 97%, Thermo Scientific™

Catalog No. AC110991000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC110991000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC110991000 Supplier Thermo Scientific Chemicals Supplier No. 110991000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1501-82-2
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00003721
InChI Key HYPABJGVBDSCIT-UPHRSURJSA-N
Synonym e-cyclododecene, cyclododecene, e, trans-cyclododecene, 1e-cyclododecene, t-cyclododecene, cyclododecene, trans, 11e-cyclododecene
PubChem CID 5365627
IUPAC Name (Z)-cyclododecene
SMILES C1CCCCC\C=C/CCCC1

Specifications

Density 0.87g/mL
Assay Percent Range 97%
Linear Formula C12H122
Quantity 100 g
Formula Weight 166.31
Chemical Name or Material Cyclododecene, 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.