accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1501-84-4

IUPAC Name: hydrogen 1-(adamantan-1-yl)ethan-1-amine chloride
Synonyms: hydrogen rimantadine chloride
Molecular Weight (g/mol): 215.77
Molecular Formula: C12H22ClN
InChi Key: OZBDFBJXRJWNAV-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)