Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Adamantyl)ethylamine hydrochloride, Thermo Scientific™

Catalog No. BTB12981EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB12981EA 10 g Amber Glass Bottle
BTB12981DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB12981EA Supplier Thermo Scientific Chemicals Supplier No. BTB12981EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1501-84-4
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00072023
InChI Key OZBDFBJXRJWNAV-UHFFFAOYNA-N
Synonym rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan
PubChem CID 15165
ChEBI CHEBI:8865
IUPAC Name 1-(1-adamantyl)ethanamine;hydrochloride
SMILES [H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 215.77
Percent Purity 95%
Chemical Name or Material 1-(1-Adamantyl)ethylamine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.