accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15015-81-3

IUPAC Name: sodium hexadecane-1-sulfonate
Synonyms: sodium hexadecane-1-sulphonate
Molecular Weight (g/mol): 328.49
Molecular Formula: C16H33NaO3S
InChi Key: PNGBYKXZVCIZRN-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCCCCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)