accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSNH2

CAS RN 150223-58-8

IUPAC Name: 4-phenylpyridine-2-carbothioamide
Synonyms: 4-phenylpyridine-2-carbothioamide
Molecular Weight (g/mol): 214.29
Molecular Formula: C12H10N2S
InChi Key: ZVJKVMYMCJFXAE-UHFFFAOYSA-N
SMILES: NC(=S)C1=NC=CC(=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results