CAS RN 15027-13-1
IUPAC Name:
2-{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms:
2-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol):
265.31
Molecular Formula:
C14H19NO4
InChi Key:
NMYWMOZOCYAHNC-UHFFFAOYNA-N
SMILES:
CCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
N-Carbobenzoxy-DL-norleucine 99.0+%, TCI America™
CAS: 15027-13-1 Molecular Formula: C14H19NO4 Molecular Weight (g/mol): 265.31 MDL Number: MFCD00037271 InChI Key: NMYWMOZOCYAHNC-UHFFFAOYNA-N Synonym: N-Cbz-DL-norleucine, Z-DL-Nle-OH PubChem CID: 263476 IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}hexanoic acid SMILES: CCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O