Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-norleucine 99.0+%, TCI America™
SDP

Catalog No. C07601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C07601G Supplier TCI America Supplier No. C07601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15027-13-1
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00037271
InChI Key NMYWMOZOCYAHNC-UHFFFAOYNA-N
Synonym N-Cbz-DL-norleucine, Z-DL-Nle-OH
PubChem CID 263476
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}hexanoic acid
SMILES CCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 105°C
Color White
Quantity 1 g
Formula Weight 265.31
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-norleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.