accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15028-41-8

IUPAC Name: methyl 2-amino-2-methylpropanoate hydrochloride
Synonyms: methyl 2-amino-2-methylpropanoate hydrochloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: NVWZNEDLYYLQJC-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C(C)(C)N
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)