Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl α-aminoisobutyrate hydrochloride, 98%, Thermo Scientific™

Catalog No. AC397980010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397980010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397980010 Supplier Thermo Scientific Chemicals Supplier No. 397980010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15028-41-8
MDL Number MFCD00214247
InChI Key NVWZNEDLYYLQJC-UHFFFAOYSA-N
Synonym methyl 2-amino-2-methylpropanoate hydrochloride, methyl 2-aminoisobutyrate hydrochloride, h-aib-ome.hcl, alpha-aminoisobutyric acid methyl ester hydrochloride, 2-aminoisobutyric acid methyl ester hydrochloride, methyl alpha-aminoisobutyrate hydrochloride, methyl 2-methylalaninate hydrochloride, alanine, 2-methyl-, methyl ester, hydrochloride, aib-ome hydrochloride, pubchem16472
PubChem CID 13258034
IUPAC Name methyl 2-amino-2-methylpropanoate;hydrochloride
SMILES CC(C)(C(=O)OC)N.Cl

Specifications

Melting Point 185.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H11NO2·HCl
Quantity 1 g
Molecular Weight (g/mol) 153.64
Formula Weight 153.64
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl α-aminoisobutyrate hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.