accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHNOOOO

CAS RN 15030-72-5

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-2-methylpropanoate
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-2-methylpropanoate
Molecular Weight (g/mol): 236.25
Molecular Formula: C12H14NO4
InChi Key: QKVCSJBBYNYZNM-UHFFFAOYSA-M
SMILES: CC(C)(NC(=O)OCC1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)