Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl (Methylthiomethyl)Phosphonate

Catalog No. 36666825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-666-825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 36-666-825G Supplier Sigma Aldrich Supplier No. 36666825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Synthesis of (+)-minfiensine via organocatalytic Diels-Alder/amine cyclization and 6-exo-dig radical cyclizationPreparation of fused nitrogen, sulfur, and phosphorus containing-heterocycles and relevant difurylpyridazine phosphonates with antimicrobial activityPreparation of phosphorus and sulfur stereoisomers of Et menthyl [(methylsulfinyl)methyl]phosphonate

Specifications

CAS 15030-72-5
Melting Point 65°C to 69°C
Molecular Formula C12H15NO4
Linear Formula C6H5CH2O2CNHC(CH3)2CO2H
Refractive Index n20/D 1.465
MDL Number MFCD00004190
Quantity 5 g
Synonym Z-2-Methylalanine
Molecular Weight (g/mol) 237.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.