accessibility menu, dialog, popup

UserName

Viewing profile as user:
OF

CAS RN 150322-73-9

IUPAC Name: 1-cyclopropyl-2-(2-fluorophenyl)ethan-1-one
Synonyms: 1-cyclopropyl-2-(2-fluorophenyl)ethanone
Molecular Weight (g/mol): 178.21
Molecular Formula: C11H11FO
InChi Key: DWBGTJUQWKWYGB-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1CC(=O)C1CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results