Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl 2-Fluorobenzyl Ketone 98.0+%, TCI America™
SDP

Catalog No. C27105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27105G 5 g
C271025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27105G Supplier TCI America Supplier No. C27105G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 150322-73-9
Molecular Formula C11H11FO
Molecular Weight (g/mol) 178.206
MDL Number MFCD10687164
InChI Key DWBGTJUQWKWYGB-UHFFFAOYSA-N
PubChem CID 15289941
IUPAC Name 1-cyclopropyl-2-(2-fluorophenyl)ethanone
SMILES C1CC1C(=O)CC2=CC=CC=C2F

Specifications

Color Yellow
Boiling Point 61°C
Quantity 5 g
Formula Weight 178.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropyl 2-Fluorobenzyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.