CAS RN 1504-06-9
IUPAC Name:
3-methyl-2,3-dihydro-1H-indol-2-one
Synonyms:
3-methyloxindole
Molecular Weight (g/mol):
147.18
Molecular Formula:
C9H9NO
InChi Key:
BBZCPUCZKLTAJQ-UHFFFAOYNA-N
SMILES:
CC1C(=O)NC2=CC=CC=C12
1
–
1
of
1
results
1
Sigma Aldrich 3-Methyl-2-oxindole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.