CAS RN 150403-89-7
IUPAC Name:
(2S)-2-azaniumyl-6-[(1-iminiumylethyl)amino]hexanoate
Synonyms:
(2S)-2-ammonio-6-[(1-iminioethyl)amino]hexanoate
Molecular Weight (g/mol):
188.25
Molecular Formula:
C8H18N3O2
InChi Key:
ONYFNWIHJBLQKE-ZETCQYMHSA-O
SMILES:
CC(=[NH2+])NCCCC[C@H]([NH3+])C([O-])=O
1
–
1
of
1
results
1
L-NIL hydrochloride, Tocris Bioscience™
CAS: 150403-89-7 Molecular Formula: C8H18ClN3O2 Molecular Weight (g/mol): 223.701 InChI Key: HJYWSATZDBEAOS-FJXQXJEOSA-N Synonym: l-nil hydrochloride,n6-1-iminoethyl-l-lysine hydrochloride,l-n 6-1-iminoethyl lysine hydrochloride,l-lysine,o-acetamidine dihydrochloride,l-n6-1-iminoethyl-lysine hydrochloride,s-n6-1-iminoethyl lysine hydrochloride PubChem CID: 9794509 IUPAC Name: (2S)-2-amino-6-(1-aminoethylideneamino)hexanoic acid;hydrochloride SMILES: CC(=NCCCCC(C(=O)O)N)N.Cl