accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNONO

CAS RN 150529-93-4

IUPAC Name: 4-phenyl-2-[2-(4-phenyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
Synonyms: 4-phenyl-2-[2-(4-phenyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 334.42
Molecular Formula: C21H22N2O2
InChi Key: JTNVCJCSECAMLD-UHFFFAOYNA-N
SMILES: CC(C)(C1=NC(CO1)C1=CC=CC=C1)C1=NC(CO1)C1=CC=CC=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results