Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-2,2'-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline]

Catalog No. AC428050010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC428050010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428050010 Supplier Thermo Scientific Chemicals Supplier No. 428050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 150529-93-4 | C21H22N2O2 | 334.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150529-93-4
Molecular Formula C21H22N2O2
Molecular Weight (g/mol) 334.42
MDL Number MFCD00192244
InChI Key JTNVCJCSECAMLD-UHFFFAOYNA-N
PubChem CID 676390
IUPAC Name (4R)-4-phenyl-2-[2-[(4R)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]propan-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C1=NC(CO1)C1=CC=CC=C1)C1=NC(CO1)C1=CC=CC=C1

Specifications

Packaging Glass bottle
Melting Point 56.0°C to 58.0°C
Merck Index 15, 1297
Quantity 1 g
Formula Weight 334.42
Chemical Name or Material (+)-2,2′-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline]

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.