accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO2H2H2H2H2H2H

CAS RN 15054-86-1

IUPAC Name: (²H₄)ethane-1,2-di(²H)ol
Synonyms: (²H₄)ethane-1,2-di(²H)ol
Molecular Weight (g/mol): 68.11
Molecular Formula: C2H6O2
InChi Key: LYCAIKOWRPUZTN-UFSLNRCZSA-N
SMILES: [2H]OC([2H])([2H])C([2H])([2H])O[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results