Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol-d{6}, 98% (Isotopic)

Catalog No. AA4228806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4228806 5 g
AA4228803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4228806 Supplier Thermo Scientific Chemicals Supplier No. 04228806
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 15054-86-1 | C2H6O2 | 68.11 g/mol

It is used in synthetic materials and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15054-86-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 68.11
MDL Number MFCD00078666
InChI Key LYCAIKOWRPUZTN-UFSLNRCZSA-N
Synonym ethylene glycol-d6, 1,1,2,2-2h4 ethane-1,2-2h2 diol, 1,2-ethane-1,1,2,2-d4-diol-d2, 2 h? ethane-1,2-di 2 h ol, ethylen-d4 glycol-d2, ethylene glycol d6, ethylene glycol-d6, 98 atom % d, ethylene glycol-d6 >99.0 atom % d
PubChem CID 2733139
IUPAC Name (²H₄)ethane-1,2-di(²H)ol
SMILES [2H]OC([2H])([2H])C([2H])([2H])O[2H]

Specifications

Physical Form Liquid
Quantity 5 g
Density 1.189 g/mL
Odor Odorless
Flash Point 111°C (232°F)
Boiling Point 86°C to 87°C (8mmHg)
Solubility Information Soluble in water,methanol,DMSO
Formula Weight 68.02
Percent Purity 98%
Chemical Name or Material Ethylene glycol-d6, Isotopic
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 239-122-6
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.