accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15060-59-0

IUPAC Name: lithium(1+) vanadium trioxidandiide
Synonyms: lithium(1+) vanadium trioxidandiide
Molecular Weight (g/mol): 105.88
Molecular Formula: LiO3V
InChi Key: IZPPMPGSWJHKSD-UHFFFAOYSA-N
SMILES: [Li+].[O--].[O--].[O--].[V]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)