Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium vanadium oxide, 99.9% (metals basis)

Catalog No. AA3935830
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3935830 250 g
AA3935818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3935830 Supplier Thermo Scientific Chemicals Supplier No. 03935830
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 15060-59-0

Lithium vanadium oxide is used in developing polymer lithium ion (PLI) batteries and also in coatings, nanowires, nanofibers, plastics, and textiles In alloy and catalyst applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15060-59-0
MDL Number MFCD00053997
Synonym Lithium vanadate

Specifications

Quantity 250 g
UN Number UN3285
Solubility Information Soluble in water
Formula Weight 105.88
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Lithium vanadium oxide
Hazard Category H302+H332-H319-H335
Hazard Statement GHS H Statement
H331-H302-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber 239-124-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.