accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CH3CH3CI

CAS RN 15066-80-5

IUPAC Name: ethyltripropylazanium iodide
Synonyms: ethyltripropylazanium iodide
Molecular Weight (g/mol): 299.24
Molecular Formula: C11H26IN
InChi Key: ZLHWTGZMAWUUMD-UHFFFAOYSA-M
SMILES: [I-].CCC[N+](CC)(CCC)CCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results