Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyltripropylammonium Iodide 99.0+%, TCI America™
SDP

Catalog No. E019125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E019125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E019125G Supplier TCI America Supplier No. E019125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15066-80-5
Molecular Formula C11H26IN
Molecular Weight (g/mol) 299.24
MDL Number MFCD00059976
InChI Key ZLHWTGZMAWUUMD-UHFFFAOYSA-M
PubChem CID 23500186
IUPAC Name ethyl(tripropyl)azanium;iodide
SMILES CCC[N+](CC)(CCC)CCC.[I-]

Specifications

Quantity 25 g
Formula Weight 299.24
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyltripropylammonium Iodide
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.