accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1510-21-0

H3CH3C(R)H3C(R)(S)(S)(S)(R)H3C(S)(R)H3COOOHO

CAS RN 1510-21-0

IUPAC Name: 4-{[(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl]oxy}-4-oxobutanoic acid
Synonyms: cholesteryl succinate
Molecular Weight (g/mol): 486.74
Molecular Formula: C31H50O4
InChi Key: WLNARFZDISHUGS-MIXBDBMTSA-N
SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)CCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results