Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cholesteryl hemisuccinate

Catalog No. 502908362
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-8362 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-8362 Supplier Sigma Aldrich Supplier No. C651225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cholesteryl hemisuccinate (CHEMS) is an acidic cholesterol ester. It self assembles into bilayers in alkaline and neutral aqueous media. CHEMS exhibits pH sensitive polymorphism. It consists of succinic acid esterified to the beta-hydroxyl group of cholesterol. This chemical modification results in the ability of CHEMS to adopt a lamellar assembly in suitable aqueous media, whereas cholesterol forms monohydrate crystals in an aqueous environment.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1510-21-0
Molecular Formula C31H50O4
Linear Formula C31H50O4
MDL Number MFCD00037705
Quantity 25 g
Synonym 3 beta-Hydroxy-5-cholestene 3-hemisuccinate; 5-Cholesten-3 beta-ol 3-hemisuccinate; Cholesteryl hydrogen succinate
Molecular Weight (g/mol) 486.73
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.