accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15100-75-1

IUPAC Name: hydrogen tert-butyl 2-amino-3-phenylpropanoate chloride
Synonyms: hydrogen tert-butyl 2-amino-3-phenylpropanoate chloride
Molecular Weight (g/mol): 257.76
Molecular Formula: C13H20ClNO2
InChi Key: FDMCEXDXULPJPG-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)(C)OC(=O)C(N)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)