accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOON

CAS RN 15103-48-7

IUPAC Name: pyridine-2-sulfonic acid
Synonyms: pyridine-2-sulfonic acid
Molecular Weight (g/mol): 159.16
Molecular Formula: C5H5NO3S
InChi Key: KZVLNAGYSAKYMG-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)