accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1510828-68-8

BOOH3CCH3H3CH3CFCH3NOO

CAS RN 1510828-68-8

IUPAC Name: 2-(2-fluoro-4-methyl-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2-fluoro-4-methyl-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 281.09
Molecular Formula: C13H17BFNO4
InChi Key: VJFYRTJYOVWHNP-UHFFFAOYSA-N
SMILES: CC1=C(C=C(B2OC(C)(C)C(C)(C)O2)C(F)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results