accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 151171-11-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-2-methylbutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-2-methylbutanoic acid
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: SHZXLTCEPXVCSV-JTQLQIEISA-N
SMILES: CCC(C)(NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)