CAS RN 151198-85-5
IUPAC Name:
2,2,2-trifluoro-N-trifluoromethanesulfonylacetamide
Synonyms:
2,2,2-trifluoro-N-trifluoromethanesulfonylacetamide
Molecular Weight (g/mol):
245.1
Molecular Formula:
C3HF6NO3S
InChi Key:
CFYBHDCZEADVJH-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(=O)NS(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
N-(Trifluoromethanesulfonyl)trifluoroacetamide 98.0+%, TCI America™
CAS: 151198-85-5 Molecular Formula: C3HF6NO3S Molecular Weight (g/mol): 245.095 InChI Key: CFYBHDCZEADVJH-UHFFFAOYSA-N PubChem CID: 12127911 IUPAC Name: 2,2,2-trifluoro-N-(trifluoromethylsulfonyl)acetamide SMILES: C(=O)(C(F)(F)F)NS(=O)(=O)C(F)(F)F