Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Trifluoromethanesulfonyl)trifluoroacetamide 98.0+%, TCI America™
SDP

Catalog No. T20881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T20881G Supplier TCI America Supplier No. T20881G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 151198-85-5
Molecular Formula C3HF6NO3S
Molecular Weight (g/mol) 245.095
InChI Key CFYBHDCZEADVJH-UHFFFAOYSA-N
PubChem CID 12127911
IUPAC Name 2,2,2-trifluoro-N-(trifluoromethylsulfonyl)acetamide
SMILES C(=O)(C(F)(F)F)NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 45°C
Quantity 1 g
Formula Weight 245.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(Trifluoromethanesulfonyl)trifluoroacetamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.