accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNF

CAS RN 1513-66-2

IUPAC Name: 2,3-difluoropyridine
Synonyms: 2,3-difluoropyridine
Molecular Weight (g/mol): 115.08
Molecular Formula: C5H3F2N
InChi Key: OGVLEPMNNPZAPS-UHFFFAOYSA-N
SMILES: FC1=CC=CN=C1F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results