Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluoropyridine 98.0+%, TCI America™
SDP

Catalog No. D38925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38925G 5 g
D38921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38925G Supplier TCI America Supplier No. D38925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1513-66-2
Molecular Formula C5H3F2N
Molecular Weight (g/mol) 115.083
MDL Number MFCD04114143
InChI Key OGVLEPMNNPZAPS-UHFFFAOYSA-N
Synonym 2,3-difluoro pyridine, difluoropyridine, pyridine, 2,3-difluoro, 2,3-difluorpyridine, 2,3-difluoropyridin, pubchem5414, 2,3-difiuoropyridine?, acmc-1bvua, 2,3-bis fluoranyl pyridine, ksc493a1t
PubChem CID 2783176
IUPAC Name 2,3-difluoropyridine
SMILES C1=CC(=C(N=C1)F)F

Specifications

Color Yellow
Boiling Point 126°C
Quantity 5 g
UN Number 1993
Formula Weight 115.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Difluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.