accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaCoOONNOOOHOHOO

CAS RN 15137-09-4

IUPAC Name: λ²-cobalt(2+) disodium hydrogen 2-[(carboxylatomethyl)({2-[(carboxylatomethyl)(hydrogen carboxylatomethyl)amino]ethyl})amino]ethanehydrogen carboxylate
Synonyms: λ²-cobalt(2+) disodium hydrogen 2-[(carboxylatomethyl)({2-[(carboxylatomethyl)(hydrogen carboxylatomethyl)amino]ethyl})amino]ethanehydrogen carboxylate
Molecular Weight (g/mol): 395.14
Molecular Formula: C10H14CoN2Na2O8
InChi Key: BVGFHSPTZBVUQN-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[Co++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([OH-])=O)CC([OH-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results